N-Ethylcyclohexylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14221-AC
L14221-AE
L14221-AE
L14221-AC
AAAL14221-AEEA
38.39
USD
CAAAAL14221-AC
AAAL14221-AE
CAAAAL14221-AE
AAAL14221-AC
N-Ethylcyclohexylamine 97%
N-Ethylcyclohexylamine
Formula:
C₈H₁₇N Boiling Pt: 163…166 °C Density: 0.850 Flash Pt: 45 °C(113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003834 CAS Number: 5459-93-8 EINECS: 226-733-8 UN: 2734 ADR: "8",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...