Borane tetrahydrofuran complex 1 mol/l in THF stabilized

Danger


Stabilization: Stabilised with sodium tetrahydridoborate 5 mmol

L13961-AE
CAAAAL13961-AE
Borane tetrahydrofuran complex 1 mol/l in THF stabilized
Borane tetrahydrofuran complex 1 mol/l in tetrahydrofuran
Formula: C₄H₁₁BO
MW: 85.94
Density: 0.878
Flash Pt: -21 °C(-8 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012429
EINECS: 237-881-8
UN: 3148
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR