1-Bromo-3-isopropoxybenzene 98%

Synonyms: 3-Bromophenyl isopropyl ether

L13802-06 L13802-14 L13802-06 L13802-14
AAAL13802-06EA 41.15 USD
CAAAAL13802-06 AAAL13802-06 AAAL13802-14 CAAAAL13802-14
1-Bromo-3-isopropoxybenzene 98%
1-Bromo-3-isopropoxybenzene
Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Boiling Pt: 222 °C
Density: 1,33
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070756
CAS Number: 131738-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR