4'-(2-Chloroethyl)acetophenone tech. 90%, Technical Grade

Warning

Synonyms: 1-[4-(2-Chloroethyl)phenyl]ethan-1-one

L13800-06 L13800-14 L13800-06 L13800-14
AAAL13800-14EA 440.67 USD
AAAL13800-14 CAAAAL13800-06 CAAAAL13800-14 AAAL13800-06
4'-(2-Chloroethyl)acetophenone tech. 90%, Technical Grade
4'-(2-Chloroethyl)acetophenone
Formula: C₁₀H₁₁ClO
MW: 182.65
Boiling Pt: 112 °C/0.1 mm
Density: 1.150
Storage Temperature: Ambient
MDL Number: MFCD00016350
CAS Number: 69614-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR