Bromopentafluoroacetone 95%

Danger

L13789-06 L13789-03 L13789-06 L13789-03
AAAL13789-03EA 47.96 USD
AAAL13789-03 CAAAAL13789-06 CAAAAL13789-03 AAAL13789-06
Bromopentafluoroacetone 95%
Bromopentafluoroacetone
Formula: C₃BrF₅O
Boiling Pt: 31…33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153080
CAS Number: 815-23-6
EINECS: 000-000-0
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR