2,4-Diaminoquinazoline 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinazoline-2,4-diamine

L13783-03 L13783-06 L13783-03 L13783-06
AAAL13783-03EA 43.09 USD
AAAL13783-03 CAAAAL13783-03 CAAAAL13783-06 AAAL13783-06
2,4-Diaminoquinazoline 98+%
2,4-Diaminoquinazoline
Formula: C₈H₈N₄
MW: 160.18 g/mol
Melting Pt: 254…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00236032
CAS Number: 1899-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR