N-(2-Bromobenzyloxycarbonyloxy)succinimide ≥98%

Warning

Synonyms: 2-Bromobenzyl Succinimidyl CarbonateCarbonic Acid 2-Bromobenzyl Succinimidyl EsterZ(2-Br)-OSu

L13471-06 L13471-14 L13471-06 L13471-14
AAAL13471-06EA 31.13 USD
CAAAAL13471-06 AAAL13471-06 CAAAAL13471-14 AAAL13471-14
N-(2-Bromobenzyloxycarbonyloxy)succinimide ≥98%
N-(2-Bromobenzyloxycarbonyloxy)succinimide
Formula: C₁₂H₁₀BrNO₅
MW: 328.12 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082848
CAS Number: 128611-93-8
UN: 3335
ADR: "9",

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR