Thiophene-3-carboxaldoxime 98%

L13114-06 L13114-03 L13114-03 L13114-06
AAAL13114-03EA 33.19 USD
CAAAAL13114-03 CAAAAL13114-06 AAAL13114-03 AAAL13114-06
Thiophene-3-carboxaldoxime 98%
Thiophene-3-carboxaldoxime
Formula: C₅H₅NOS
Melting Pt: 121…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00151853
CAS Number: 42466-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR