2-Indanyl-p-toluenesulfonate ≥98%

L12916-06 L12916-14 L12916-06 L12916-14
AAAL12916-06EA 36.33 USD
AAAL12916-06 CAAAAL12916-14 AAAL12916-14 CAAAAL12916-06
2-Indanyl-p-toluenesulfonate ≥98%
2-Indanyl-p-toluenesulfonate
Formula: C₁₆H₁₆O₃S
MW: 288.37 g/mol
Melting Pt: 118…121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082747
CAS Number: 17783-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR