Allyl ethyl sulfide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12847-06
L12847-06
AAAL12847-06EA
105.99
USD
CAAAAL12847-06
AAAL12847-06
Allyl ethyl sulfide 97%
Allyl ethyl sulphide
Formula:
C₅H₁₀S Boiling Pt: 115…116 °C Density: 0.868 Flash Pt: 18 °C(64 °F) Storage Temperature: Ambient |
UN:
1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...