3-Ethylamino-p-cresol 90%, tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Ethylamino-4-methylphenol

L12768-06 L12768-06
AAAL12768-06EA 47.69 USD
CAAAAL12768-06 AAAL12768-06
3-Ethylamino-p-cresol 90%, tech.
3-Ethylamino-p-cresol
Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 135…140 °C/3 mm
Melting Pt: 84…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00002382
CAS Number: 120-37-6
EINECS: 204-391-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR