3-(4-Methylsulfonylbenzoyl)propionic acid 98%

Warning

L12750-06 L12750-03 L12750-03 L12750-06
AAAL12750-03EA 61.89 USD
CAAAAL12750-03 AAAL12750-03 CAAAAL12750-06 AAAL12750-06
3-(4-Methylsulfonylbenzoyl)propionic acid 98%
3-(4-Methylsulfonylbenzoyl)propionic acid
Formula: C₁₁H₁₂O₅S
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00082738
CAS Number: 7028-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR