4-(4-Chlorophenyl)-3-thiosemicarbazide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-(4-Chlorophenyl)hydrazinecarbothioamide
L12657-06
L12657-06
AAAL12657-06EA
163.99
USD
AAAL12657-06
CAAAAL12657-06
4-(4-Chlorophenyl)-3-thiosemicarbazide 97%
4-(4-Chlorophenyl)-3-thiosemicarbazide
Formula:
C₇H₈ClN₃S MW: 201.68 g/mol Melting Pt: ca 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00041295 CAS Number: 22814-92-2 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...