1-Fluoro-4-(trifluoromethylthio)benzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluorophenyl trifluoromethyl sulphide

L12485-03 L12485-03
AAAL12485-03EA 70.39 USD
AAAL12485-03 CAAAAL12485-03
1-Fluoro-4-(trifluoromethylthio)benzene 98+%
1-Fluoro-4-(trifluoromethylthio)benzene
Formula: C₇H₄F₄S
MW: 196.17
Boiling Pt: 136…138 °C
Density: 1.38
Flash Pt: 37 °C(99 °F)
Storage Temperature: Ambient
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR