1-Piperidinethiocarboxamide 98%

Warning

L12445-03 L12445-06 L12445-06 L12445-03
AAAL12445-03EA 67.36 USD
AAAL12445-03 CAAAAL12445-03 CAAAAL12445-06 AAAL12445-06
1-Piperidinethiocarboxamide 98%
1-Piperidinethiocarboxamide
Formula: C₆H₁₂N₂S
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00041230
CAS Number: 14294-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR