4-(Diethylamino)phenyl isothiocyanate 97%

Danger

L12420-06 L12420-06 L12420-03 L12420-03
AAAL12420-06EA 116.55 USD
AAAL12420-06 CAAAAL12420-06 AAAL12420-03 CAAAAL12420-03
4-(Diethylamino)phenyl isothiocyanate 97%
4-(Diethylamino)phenyl isothiocyanate
Formula: C₁₁H₁₄N₂S
Boiling Pt: 147…148 °C/1.2 mm
Melting Pt: 30…33 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041094
CAS Number: 84381-54-4
EINECS: 000-000-0
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR