(R)-(-)-1-(1-Naphthyl)ethyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12286-MD
L12286-03
L12286-MD
L12286-03
AAAL12286-MDEA
41.99
USD
AAAL12286-MD
AAAL12286-03
CAAAAL12286-MD
CAAAAL12286-03
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate 97%
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate
Formula:
C₁₃H₁₁NO Boiling Pt: 106…108°/0.16m Density: 1.118 Flash Pt: 93 °C(199 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004015 CAS Number: 42340-98-7 EINECS: 255-759-2 UN: 2206 ADR: "6.1",III |
Specification Test Results
Optical rotation | -47° (c=3.5 in toluene) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...