Diethyl-sec-butylmalonate 96%

Synonyms: Diethyl 2-(sec-butyl)malonate

L12253-06 L12253-14 L12253-14 L12253-06
AAAL12253-14EA 149.96 USD
CAAAAL12253-14 CAAAAL12253-06 AAAL12253-14 AAAL12253-06
Diethyl-sec-butylmalonate 96%
Diethyl sec-butylmalonate
Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 110…114 °C/18 mm
Density: 0.980
Storage Temperature: Ambient
EINECS: 201-463-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR