(S)-(+)-3-Aminopyrrolidine dihydrochloride 98%
|
Warning
|
Synonyms:
(S)-Pyrrolidin-3-amine dihydrochloride
L12222-MD
L12222-03
L12222-03
L12222-MD
AAAL12222-MDEA
37.39
USD
AAAL12222-MD
CAAAAL12222-MD
AAAL12222-03
CAAAAL12222-03
(S)-(+)-3-Aminopyrrolidine dihydrochloride 98%
(S)-(+)-3-Aminopyrrolidine dihydrochloride
Formula:
C₄H₁₂Cl₂N₂ MW: 159.06 g/mol Melting Pt: ca 283 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00070605 CAS Number: 116183-83-6 |
Specification Test Results
Optical rotation | +1.2° (c=10 in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...