Propargyl p-toluenesulfonate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Propargyl p-toluenesulfonatePropargyl p-toluenesulphonate

L12075-09 L12075-09
AAAL12075-09EA 79.09 USD
AAAL12075-09 CAAAAL12075-09
Propargyl p-toluenesulfonate 97%
Propargyl p-toluenesulfonate
Formula: C₁₀H₁₀O₃S
MW: 210.25 g/mol
Boiling Pt: 117…121 °C/0.3 mm
Density: 1.220
Storage Temperature: Ambient
MDL Number: MFCD00078365
CAS Number: 6165-76-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR