2-(4-Morpholino)ethyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals

Danger

L12046-04 L12046-04
AAAL12046-04EA 69.6 USD
AAAL12046-04 CAAAAL12046-04
2-(4-Morpholino)ethyl isothiocyanate 97%
2-(4-Morpholino)ethyl isothiocyanate
Formula: C₇H₁₂N₂OS
Boiling Pt: 162…163 °C/10 mm
Density: 1.110
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00041221
CAS Number: 63224-35-1
EINECS: 000-000-0
UN: 2922
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR