1H,1H,2H,2H-Heptadecafluorodecyl acrylate 96% stabilized

Warning

Synonyms: Acrylic Acid 1H,1H,2H,2H-Heptadecafluorodecyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 100 ppm

L11993-06 L11993-06
AAAL11993-06EA 69.2 USD
CAAAAL11993-06 AAAL11993-06
1H,1H,2H,2H-Heptadecafluorodecyl acrylate 96% stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate
Formula: C₁₃H₇F₁₇O₂
MW: 518.17 g/mol
Boiling Pt: 89…90 °C/4 mm
Melting Pt: –3 °C
Density: 1.637
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042306
CAS Number: 27905-45-9
EINECS: 248-722-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR