Piperonaldoxime 99%

L11952-06 L11952-14 L11952-14 L11952-06
AAAL11952-14EA 200.91 USD
AAAL11952-14 CAAAAL11952-06 CAAAAL11952-14 AAAL11952-06
Piperonaldoxime 99%
Piperonaldoxime
Formula: C₈H₇NO₃
MW: 165.15 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00016905
CAS Number: 2089-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR