Cyclopropylacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Cyclopropylacetonitrile

L11950-14 L11950-06 L11950-14 L11950-06
AAAL11950-14EA 273.99 USD
CAAAAL11950-14 AAAL11950-14 AAAL11950-06 CAAAAL11950-06
Cyclopropylacetonitrile 97%
Cyclopropylacetonitrile
Formula: C₅H₇N
MW: 81.12 g/mol
Boiling Pt: 142…144 °C
Density: 0.890
Flash Pt: 43 °C(109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041523
CAS Number: 6542-60-5
UN: 3275
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR