4-Bromo-1,3,5-trimethyl-1H-pyrazole 95%
|
Warning
|
Synonyms:
4-Bromo-1,3,5-trimethylpyrazole
L11876-03
L11876-06
L11876-03
L11876-06
AAAL11876-03EA
30.04
USD
AAAL11876-03
AAAL11876-06
CAAAAL11876-06
CAAAAL11876-03
4-Bromo-1,3,5-trimethyl-1H-pyrazole 95%
4-Bromo-1,3,5-trimethyl-1H-pyrazole
Formula:
C₆H₉BrN₂ MW: 189.06 Boiling Pt: 101…102 °C/15 mm Melting Pt: 32…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052532 CAS Number: 15801-69-1 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...