2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate 98%
|
Warning
|
L11817-06
L11817-06
L11817-03
L11817-03
AAAL11817-03EA
63.9
USD
CAAAAL11817-03
AAAL11817-03
CAAAAL11817-06
AAAL11817-06
2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate 98%
2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate
Formula:
C₁₁H₁₃NO₂S Boiling Pt: 150…151 °C/0.3 mm Density: 1.170 Storage Temperature: Ambient |
MDL Number:
MFCD00041366 CAS Number: 21714-25-0 EINECS: 000-000-0 UN: 2922 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...