(-)-cis-2-Benzylaminocyclohexanemethanol 98%
|
Warning
|
Synonyms:
(-)-N-Benzyl-cis-2-hydroxymethylcyclohexylamine
L11749-03
L11749-03
L11749-MD
L11749-MD
AAAL11749-03EA
81.03
USD
AAAL11749-03
CAAAAL11749-MD
AAAL11749-MD
CAAAAL11749-03
(-)-cis-2-Benzylaminocyclohexanemethanol 98%
(-)-cis-2-Benzylaminocyclohexanemethanol
Formula:
C₁₄H₂₁NO MW: 219.33 g/mol Melting Pt: 66…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151396 CAS Number: 71581-93-6 |
Specification Test Results
Optical rotation | -24° (c=1 in methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...