1,2-Dimethoxy-4,5-dinitrobenzene 96%

Supplier: Thermo Scientific Chemicals

Warning

L11706-06 L11706-06
AAAL11706-06EA 195 USD
AAAL11706-06 CAAAAL11706-06
1,2-Dimethoxy-4,5-dinitrobenzene 96%
1,2-Dimethoxy-4,5-dinitrobenzene
Formula: C₈H₈N₂O₆
Melting Pt: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00082695
CAS Number: 3395-03-7
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR