N-(2-Cyanoethyl)morpholine ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Morpholino)propionitrile3-(morpholin-4-yl)propanenitrile3-Morpholin-4-ylpropanenitrile3-Morpholinopropionitrile

L11664-14 L11664-14
AAAL11664-14EA 93.09 USD
CAAAAL11664-14 AAAL11664-14
N-(2-Cyanoethyl)morpholine ≥98%
N-(2-Cyanoethyl)morpholine
Formula: C₇H₁₂N₂O
MW: 140.19 g/mol
Boiling Pt: 106…109 °C/7 mm
Melting Pt: 20…21 °C
Density: 1.037
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006178
CAS Number: 4542-47-6
EINECS: 224-894-9
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR