4-(3-Fluorophenyl)-3-thiosemicarbazide 97%
|
Danger
|
L11652-03
L11652-06
L11652-03
L11652-06
AAAL11652-03EA
51.84
USD
CAAAAL11652-03
AAAL11652-03
AAAL11652-06
CAAAAL11652-06
4-(3-Fluorophenyl)-3-thiosemicarbazide 97%
4-(3-Fluorophenyl)-3-thiosemicarbazide
Formula:
C₇H₈FN₃S Melting Pt: ca 160 °C (dec.) Storage Temperature: Ambient |
UN:
2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...