3-Di-n-propylamino-1-propyne 98%

Danger

L11633-06 L11633-03 L11633-03 L11633-06
AAAL11633-06EA 95.85 USD
AAAL11633-06 CAAAAL11633-03 CAAAAL11633-06 AAAL11633-03
3-Di-n-propylamino-1-propyne 98%
3-Di-n-propylamino-1-propyne
Formula: C₉H₁₇N
MW: 139.24 g/mol
Boiling Pt: 157…159 °C
Density: 0.828
Flash Pt: 55 °C(131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041626
CAS Number: 6323-79-1
UN: 2733
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR