4-(3-Chlorophenyl)-3-thiosemicarbazide 98+%
|
Danger
|
L11610-03
L11610-06
L11610-03
L11610-06
AAAL11610-06EA
163.01
USD
AAAL11610-06
CAAAAL11610-03
AAAL11610-03
CAAAAL11610-06
4-(3-Chlorophenyl)-3-thiosemicarbazide 98+%
4-(3-Chlorophenyl)-3-thiosemicarbazide
Formula:
C₇H₈ClN₃S Melting Pt: 130…133 °C Storage Temperature: Ambient |
UN:
2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...