trans-2-Undecenoic acid ≥80%, tech.

Warning

Synonyms: (E)-Undec-2-enoic acid

L11579-06 L11579-06 L11579-03 L11579-03
AAAL11579-03EA 41.89 USD
CAAAAL11579-03 AAAL11579-03 CAAAAL11579-06 AAAL11579-06
trans-2-Undecenoic acid ≥80%, tech.
trans-2-Undecenoic acid
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 138…140 °C/2 mm
Storage Temperature: Ambient
MDL Number: MFCD00040909
CAS Number: 15790-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR