3-Amino-4-(methylthio)benzotrifluoride 97%
|
Warning
|
Synonyms:
2-Methylthio-5-(trifluoromethyl)aniline
L11452-03
L11452-03
AAAL11452-03EA
83.03
USD
CAAAAL11452-03
AAAL11452-03
3-Amino-4-(methylthio)benzotrifluoride 97%
3-Amino-4-(methylthio)benzotrifluoride
Formula:
C₈H₈F₃NS Boiling Pt: 111…112 °C/12 mm Storage Temperature: Ambient |
MDL Number:
MFCD00052715 UN: 2810 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...