4-Chlorocinnamamide 97%

Supplier: Thermo Scientific Chemicals

L11406-03 L11406-03 L11406-06 L11406-06
AAAL11406-06EA 239.99 USD
AAAL11406-06 AAAL11406-03 CAAAAL11406-03 CAAAAL11406-06
4-Chlorocinnamamide 97%
4-Chlorocinnamamide
Formula: C₉H₈ClNO
MW: 181.62 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00017147
CAS Number: 18166-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR