3-Butoxypropionitrile 97%

Warning

Synonyms: 3-n-Butoxypropionitrile3-Butoxypropanenitrile

L11342-06 L11342-14 L11342-06 L11342-14
AAAL11342-14EA 101.82 USD
CAAAAL11342-14 CAAAAL11342-06 AAAL11342-14 AAAL11342-06
3-Butoxypropionitrile 97%
3-Butoxypropionitrile
Formula: C₇H₁₃NO
MW: 127.19 g/mol
Boiling Pt: 105…106 °C/15 mm
Density: 0,920
Flash Pt: 89 °C(192 °F)
Storage Temperature: Ambient
EINECS: 230-153-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR