Benzoylurea 97%

Synonyms: Benzoylcarbamide

L11313-14 L11313-06 L11313-06 L11313-14
AAAL11313-06EA 26.25 USD
AAAL11313-06 CAAAAL11313-06 AAAL11313-14 CAAAAL11313-14
Benzoylurea 97%
Benzoylurea
Formula: C₈H₈N₂O₂
MW: 164.16 g/mol
Melting Pt: 209…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00025441
CAS Number: 614-22-2
EINECS: 210-372-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR