6-Fluoro-4-chromanone 97%
L11209-03
L11209-06
AAAL11209-06EA
127.84
USD
AAAL11209-06
AAAL11209-03
6-Fluoro-4-chromanone 97%
6-Fluoro-4-chromanone
Formula:
C₉H₇FO₂ MW: 166.15 g/mol Melting Pt: 114…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038654 CAS Number: 66892-34-0 EINECS: 266-511-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...