3,5-Dimethoxy-4-hydroxybenzhydrazide 97%
|
Warning
|
Synonyms:
4-Hydroxy-3,5-dimethoxybenzhydrazide
, 4-Hydroxy-3,5-dimethoxybenzohydrazide
L11032-06
L11032-06
L11032-03
L11032-03
AAAL11032-03EA
49.43
USD
CAAAAL11032-06
AAAL11032-03
AAAL11032-06
CAAAAL11032-03
3,5-Dimethoxy-4-hydroxybenzhydrazide 97%
3,5-Dimethoxy-4-hydroxybenzhydrazide
Formula:
C₉H₁₂N₂O₄ MW: 212.21 g/mol Melting Pt: 154…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017077 CAS Number: 1443-76-1 EINECS: 215-884-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...