4-tert-Pentylbenzenesulfonyl chloride 97%

Danger

L10894-06 L10894-06
AAAL10894-06EA 112.33 USD
AAAL10894-06 CAAAAL10894-06
4-tert-Pentylbenzenesulfonyl chloride 97%
4-tert-Pentylbenzenesulfonyl chloride
Formula: C₁₁H₁₅ClO₂S
MW: 246.76 g/mol
Melting Pt: 44…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00041509
CAS Number: 169677-20-7
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR