N-Cyclohexylethanolamine 97%
|
Warning
|
Synonyms:
N-(2-Hydroxyethyl)cyclohexylamine
, 2-(Cyclohexylamino)ethanol
L10821-14
L10821-22
L10821-14
AAAL10821-14EA
58.4
USD
CAAAAL10821-22
AAAL10821-14
CAAAAL10821-14
N-Cyclohexylethanolamine 97%
N-Cyclohexylethanolamine
Formula:
C₈H₁₇NO MW: 143.23 g/mol Boiling Pt: 112 °C/3 mm Melting Pt: 33…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021326 CAS Number: 2842-38-8 EINECS: 220-636-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...