9-Acridinecarboxylic acid hydrate 97%
|
Warning
|
Synonyms:
Acridine-9-carboxylic acid hydrate
L10797-06
L10797-06
L10797-03
L10797-03
AAAL10797-06EA
270.89
USD
AAAL10797-06
CAAAAL10797-06
AAAL10797-03
CAAAAL10797-03
9-Acridinecarboxylic acid hydrate 97%
9-Acridinecarboxylic acid hydrate
Formula:
C₁₄H₁₁NO₃ MW: 241,24 g/mol Melting Pt: ca 290 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149578 CAS Number: 332927-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...