5-Nitrothiophene-3-carbaldehyde 97%
L10793-06
L10793-03
L10793-06
L10793-03
AAAL10793-06EA
189.53
USD
AAAL10793-06
AAAL10793-03
CAAAAL10793-06
CAAAAL10793-03
5-Nitrothiophene-3-carbaldehyde 97%
5-Nitrothiophene-3-carbaldehyde
Formula:
C₅H₃NO₃S MW: 157.15 g/mol Melting Pt: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052581 CAS Number: 75428-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...