2-Methoxyethyl phenyl sulfide 98%
L10464-06
L10464-06
L10464-14
L10464-14
AAAL10464-14EA
104.67
USD
CAAAAL10464-14
CAAAAL10464-06
AAAL10464-14
AAAL10464-06
2-Methoxyethyl phenyl sulfide 98%
2-Methoxyethyl phenyl sulphide
Formula:
C₉H₁₂OS Boiling Pt: 125…126 °C/18 mm Storage Temperature: Ambient |
MDL Number:
MFCD00041329 CAS Number: 77298-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...