1H,1H-Perfluorooctyl methacrylate 97%

Warning

L10440-03 L10440-06 L10440-06 L10440-03
AAAL10440-06EA 108.97 USD
AAAL10440-06 CAAAAL10440-06 AAAL10440-03 CAAAAL10440-03
1H,1H-Perfluorooctyl methacrylate 97%
1H,1H-Perfluorooctyl methacrylate
Formula: C₁₂H₇F₁₅O₂
Boiling Pt: 70 °C/1 mm
Density: 1.496
Flash Pt: 96 °C(204 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00039247
CAS Number: 3934-23-4
EINECS: 223-509-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR