10-Phenyl-1-decanol 96%

L10269-06 L10269-03 L10269-03 L10269-06
AAAL10269-06EA 343.84 USD
AAAL10269-06 CAAAAL10269-06 AAAL10269-03 CAAAAL10269-03
10-Phenyl-1-decanol 96%
10-Phenyl-1-decanol
Formula: C₆H₅(CH₂)₁₀OH
Boiling Pt: 145…147 °C/0.5 mm
Melting Pt: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00014426
CAS Number: 62607-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR