4-(Benzyloxy)phenyl isothiocyanate 98%

Danger

L10038-06 L10038-03 L10038-06 L10038-03
AAAL10038-06EA 233.38 USD
AAAL10038-06 CAAAAL10038-03 AAAL10038-03 CAAAAL10038-06
4-(Benzyloxy)phenyl isothiocyanate 98%
4-(Benzyloxy)phenyl isothiocyanate
Formula: C₁₄H₁₁NOS
MW: 241.31
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00041095
CAS Number: 139768-71-1
EINECS: 000-000-0
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR