Ethyl-2-isocyanato-3-methylbutyrate 97%

Warning

L09988-04 L09988-09 L09988-09 L09988-04
AAAL09988-09EA 209.79 USD
AAAL09988-09 CAAAAL09988-04 CAAAAL09988-09 AAAL09988-04
Ethyl-2-isocyanato-3-methylbutyrate 97%
Ethyl-2-isocyanato-3-methylbutyrate
Formula: C₈H₁₃NO₃
MW: 171.20 g/mol
Boiling Pt: 81…82 °C/3 mm
Storage Temperature: Refrigerator
MDL Number: MFCD00078478
CAS Number: 5296-78-6
UN: 2206
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR