Hexyl acrylate 95% stabilized

Warning

Synonyms: Acrylic Acid Hexyl Ester

Stabilization: Stabilised with hydroquinone (HQ)

L09905-14 L09905-14 L09905-22 L09905-22
AAAL09905-22EA 81.25 USD
CAAAAL09905-22 AAAL09905-22 CAAAAL09905-14 AAAL09905-14
Hexyl acrylate 95% stabilized
Hexyl acrylate
Formula: C₉H₁₆O₂
MW: 156.23 g/mol
Boiling Pt: 88…89 °C/24 mm
Melting Pt: –45 °C
Density: 0.890
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048881
CAS Number: 2499-95-8
EINECS: 219-698-5
UN: 3082
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR