Diethyl dibutylmalonate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Diethyl 2,2-dibutylmalonate

L09902-06 L09902-06 L09902-14 L09902-14
AAAL09902-06EA 35.08 USD
AAAL09902-06 CAAAAL09902-06 AAAL09902-14 CAAAAL09902-14
Diethyl dibutylmalonate 98+%
Diethyl dibutylmalonate
Formula: C₁₅H₂₈O₄
MW: 272.39 g/mol
Boiling Pt: 150 °C/12 mm
Density: 0,950
Storage Temperature: Ambient
MDL Number: MFCD00026842
EINECS: 209-890-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR